missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-Octopamine hydrochloride, 99%

alpha-adrenoceptor agonist; invertebrate neurotransmitter and neuromodulator

Manufacturer:  Alfa Aesar™ J6128106

 View more versions of this product

Catalog No. AAJ6128106



Description

Description

Applications
Octopamine HCL has the ability to halt catabolism of proteinaceous tissues. It is very important while losing weight to maximize adipose loss while minimizing the loss of muscle and other lean body tissue. Octopamine Hcl (norepinephrine) is best known as an effective ephedrine alternative, claiming to boost energy while reduce weight in bodybuilding industry for weight loss purpose. In addition, many users believe octopamine hcl is a good nootropic compound that makes our brain easy to focus, enhances motivation as a wakefulness promoter.

Solubility
It is soluble in water.

Notes
Handle under argon. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

(+/-)-Octopamine hydrochloride
170°C (decomposition)
770-05-8
MFCD00012881
14,6759
It is soluble in water.
C1=CC(=CC=C1C(CN)O)O.Cl
189.639
189.64
99%
White
5g
C8H12ClNO2
3915414
+,--octopamine hcl, +,--octopamine hydrochloride, +/--octopamine hydrochloride, 4-2-amino-1-hydroxyethyl phenol hydrochloride, 4-2-amino-1-hydroxyethyl phenol;hydrochloride, dl-octopamine hydrochloride, epirenor, octopamine hcl, octopamine hydrochloride
PUMZXCBVHLCWQG-UHFFFAOYSA-N
4-(2-amino-1-hydroxyethyl)phenol;hydrochloride
102484
Powder

Chemical Identifiers

770-05-8
189.639
PUMZXCBVHLCWQG-UHFFFAOYSA-N
102484
C1=CC(=CC=C1C(CN)O)O.Cl
C8H12ClNO2
MFCD00012881
+,--octopamine hcl, +,--octopamine hydrochloride, +/--octopamine hydrochloride, 4-2-amino-1-hydroxyethyl phenol hydrochloride, 4-2-amino-1-hydroxyethyl phenol;hydrochloride, dl-octopamine hydrochloride, epirenor, octopamine hcl, octopamine hydrochloride
4-(2-amino-1-hydroxyethyl)phenol;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 212-216-4

TSCA : No

Recommended Storage : Ambient temperatures

Documents