missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(±)-Octopamine hydrochloride, 95%, Acros Organics™

Manufacturer:  Acros Organics AC157200010

 View more versions of this product

Catalog No. AC157200010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(±)-Octopamine hydrochloride
1% max.
1g
C8H12ClNO2
octopamine hydrochloride, dl-octopamine hydrochloride, octopamine hcl, epirenor, +/--octopamine hydrochloride, +,--octopamine hcl, octopamine hydrochloride, 4-2-amino-1-hydroxyethyl phenol hydrochloride, +,--octopamine hydrochloride, 4-2-amino-1-hydroxyethyl phenol;hydrochloride
C1=CC(=CC=C1C(CN)O)O.Cl
189.639
189.64
Authentic
169.0°C to 171.0°C
770-05-8
HOC6H4CH(CH2NH2)OH·HCl
PUMZXCBVHLCWQG-UHFFFAOYSA-N
4-(2-amino-1-hydroxyethyl)phenol;hydrochloride
102484
95%

Chemical Identifiers

770-05-8
189.639
octopamine hydrochloride, dl-octopamine hydrochloride, octopamine hcl, epirenor, +/--octopamine hydrochloride, +,--octopamine hcl, octopamine hydrochloride, 4-2-amino-1-hydroxyethyl phenol hydrochloride, +,--octopamine hydrochloride, 4-2-amino-1-hydroxyethyl phenol;hydrochloride
4-(2-amino-1-hydroxyethyl)phenol;hydrochloride
C8H12ClNO2
PUMZXCBVHLCWQG-UHFFFAOYSA-N
102484
C1=CC(=CC=C1C(CN)O)O.Cl
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

Documents