Please login to your online account to display your discounted pricing

OctMAB, Tocris™

Dynamin inhibitor

$76.90 - $293.30

Chemical Identifiers

Chemical Name or Material OctMAB
Assay Percent Range >98%
Molecular Formula C21H46BrN
Formula Weight 392.5
Synonym Octadecyltrimethylammonium bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

422510

 
r & d systems
RDS422510
10mg Each for $76.90

422550

 
r & d systems
RDS422550
50mg Each for $293.30
Description & Specifications

Specifications

Formula Weight 392.5
Quantity 10mg
Chemical Name or Material OctMAB
CAS 1120-02-1
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >98%
Molecular Formula C21H46BrN
Synonym Octadecyltrimethylammonium bromide
Safety and Handling

Desiccate at Room Temperature