missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Octenidine dihydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6225706
Description
Octenidine dihydrochloride is used as a antiseptic for skin, mucous Membranes and wounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsOctenidine dihydrochloride is used as a antiseptic for skin, mucous Membranes and wounds.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Octenidine dihydrochloride | |
214°C | |
MFCD01938808 | |
14,6754 | |
Soluble in dimethyl sulfoxide and methanol. | |
98% |
70775-75-6 | |
C36H64Cl2N4 | |
5 g | |
N,N'-[Decane-1,10-diyldi-1(4H)-pyridyl-4-ylidene]bis(octylammonium) dichloride | |
623.83 |
Chemical Identifiers
70775-75-6 | |
C36H64Cl2N4 | |
MFCD01938808 | |
N,N'-[Decane-1,10-diyldi-1(4H)-pyridyl-4-ylidene]bis(octylammonium) dichloride |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 274-861-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only