missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
gamma-Octanoic lactone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 129400250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
γ-Octanoic lactone | |
104-50-7 | |
0.9800g/mL | |
>110°C | |
97.5% min. (GC) | |
C8H14O2 | |
MFCD00005402 | |
0.98 | |
IPBFYZQJXZJBFQ-UHFFFAOYSA-N | |
5-butyloxolan-2-one | |
7704 | |
98% |
98% | |
Colorless | |
234.0°C | |
Authentic | |
Glass bottle | |
1.4430 to 1.4450 | |
25 g | |
gamma-octalactone, 4-octanolide, 5-butyldihydrofuran-2 3h-one, octanolide-1,4, 2 3h-furanone, 5-butyldihydro, octan-4-olide, gamma-octanolactone, 8-oxo-5-octanolide, gamma-octanoic lactone, 5-butyltetrahydro-2-furanone | |
CCCCC1CCC(=O)O1 | |
142.2 | |
142.2 | |
Liquid |
Chemical Identifiers
104-50-7 | |
142.2 | |
IPBFYZQJXZJBFQ-UHFFFAOYSA-N | |
7704 | |
CCCCC1CCC(=O)O1 |
C8H14O2 | |
MFCD00005402 | |
gamma-octalactone, 4-octanolide, 5-butyldihydrofuran-2 3h-one, octanolide-1,4, 2 3h-furanone, 5-butyldihydro, octan-4-olide, gamma-octanolactone, 8-oxo-5-octanolide, gamma-octanoic lactone, 5-butyltetrahydro-2-furanone | |
5-butyloxolan-2-one |
Safety and Handling
EINECSNumber : 203-208-1
RTECSNumber : LU3562000
TSCA : TSCA
RUO – Research Use Only