missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

γ-Octanoic Lactone 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 129401000

 View more versions of this product

Catalog No. AC129401000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

γ-Octanoic lactone
104-50-7
Authentic
Liquid
98%
C8H14O2
MFCD00005402
100g
98%
0.98g/mL
Glass bottle
≥97.5%
1.4430 to 1.4450 (20°C, 589nm)
142.2
Clear colorless

Chemical Identifiers

104-50-7
C8H14O2
MFCD00005402
≥97.5%
98%
142.2
γ-Octanoic lactone, 98%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 203-208-1

RTECSNumber : LU3562000

TSCA : TSCA