Please login to your online account to display your discounted pricing

Octanesulfonyl chloride, 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  353600050

 View more versions of this product

Catalog No. AC353600050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid
Formula Weight 212.73
Quantity 5g
Color Clear colorless to slightly yellow
EINECS Number 232-249-8
TSCA TSCA
Density 1.0870g/mL
CAS Min % 95.0
CAS Max % 100.0
Refractive Index 1.4580 to 1.4610 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula CH3(CH2)7SO2Cl
Assay Percent Range 96%
MDL Number MFCD00014729
Name Note 96%
Chemical Name or Material Octanesulfonyl chloride
Molecular Formula C8H17ClO2S
CAS 7795-95-1

Specifications

Chemical Name or Material Octanesulfonyl chloride
Name Note 96%
CAS 7795-95-1
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C8H17ClO2S
Linear Formula CH3(CH2)7SO2Cl
Formula Weight 212.73
MDL Number MFCD00014729
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents