missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octamethylcyclotetrasiloxane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 216470250

 View more versions of this product

Catalog No. AC216470250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Octamethylcyclotetrasiloxane
97.5
17.0°C to 18.0°C
0.9560g/mL
56°C
98%
C8H24O4Si4
MFCD00003269
04, III, 1885
15, 6836
Solubility in water: insoluble. Other solubilities: soluble in carbon tetrachloride
C[Si]1(O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)C
296.61
CHEBI:25640
98%
556-67-2
100.0
Colorless
175.0°C to 176.0°C
Authentic
Glass bottle
1.3950 to 1.3970
25 g
0.956
octamethylcyclotetrasiloxane, cyclotetrasiloxane, octamethyl, oktamethylcyklotetrasiloxan, cyclic dimethylsiloxane tetramer, omcts, nuc silicone vs 7207, union carbide 7207, silicone sf 1173, oktamethylzyklotetrasiloxan, unii-cz227117je
HMMGMWAXVFQUOA-UHFFFAOYSA-N
2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
11169
296.61
Liquid

Chemical Identifiers

556-67-2
296.61
HMMGMWAXVFQUOA-UHFFFAOYSA-N
11169
2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
C8H24O4Si4
MFCD00003269
octamethylcyclotetrasiloxane, cyclotetrasiloxane, octamethyl, oktamethylcyklotetrasiloxan, cyclic dimethylsiloxane tetramer, omcts, nuc silicone vs 7207, union carbide 7207, silicone sf 1173, oktamethylzyklotetrasiloxan, unii-cz227117je
CHEBI:25640
C[Si]1(O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)C
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of damaging fertility.
May cause long lasting harmful effects to aquatic life.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/at

GHS Signal Word: Warning

EINECSNumber : 209-136-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only