missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Octamethylcyclotetrasilazane, 97%

Manufacturer:  Alfa Aesar™ L1656314

 View more versions of this product

Catalog No. AAL1656314



Specifications

Specifications

Octamethylcyclotetrasilazane
C8H28N4Si4
UN2923
292.68
Ammonia-like
94°C to 98°C
66°C (150°F)
1020-84-4
MFCD00046123
976248
97%
224°C to 225°C
25g

Chemical Identifiers

1020-84-4
C8H28N4Si4
MFCD00046123
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

TSCA : TSCA

Documents