missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Octakis(trimethylsiloxy)silsesquioxane

Manufacturer:  Alfa Aesar™ 4423103

 View more versions of this product

Catalog No. AA4423103



Specifications

Specifications

Octakis (trimethylsiloxy)silsesquioxane
1g
C24H72O10Si11
octakis trimethylsiloxy si-lsesquioxane
VLEKPQBXDHFSQO-UHFFFAOYSA-N
bis(trimethylsilyl) bis[tris(trimethylsilyloxy)silyl] silicate
71306897
Powder
White
51777-38-9
MFCD01310212
Sparingly Soluble (0.61g/L) (25°C),Calc.
C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
829.765
1130.18

Chemical Identifiers

51777-38-9
829.765
VLEKPQBXDHFSQO-UHFFFAOYSA-N
71306897
C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
C24H72O10Si11
MFCD01310212
octakis trimethylsiloxy si-lsesquioxane
bis(trimethylsilyl) bis[tris(trimethylsilyloxy)silyl] silicate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents