Learn More
Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04423122
Description
Used as flavor and fragrance intermediates. It is an important raw material used intermediate in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Octakis (trimethylsiloxy)silsesquioxane | |
White | |
MFCD01310212 | |
octakis trimethylsiloxy si-lsesquioxane, octakis trimethylsiloxy-t8-silsesquioxane | |
VLEKPQBXDHFSQO-UHFFFAOYSA-N | |
bis(trimethylsilyl) bis[tris(trimethylsilyloxy)silyl] silicate | |
71306897 | |
Powder |
51777-38-9 | |
C24H72O10Si11 | |
100 g | |
Sparingly Soluble (0.61g/L) (25°C),Calc. | |
C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C | |
829.765 | |
1130.18 |
Chemical Identifiers
51777-38-9 | |
829.765 | |
VLEKPQBXDHFSQO-UHFFFAOYSA-N | |
71306897 | |
C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C |
C24H72O10Si11 | |
MFCD01310212 | |
octakis trimethylsiloxy si-lsesquioxane, octakis trimethylsiloxy-t8-silsesquioxane | |
bis(trimethylsilyl) bis[tris(trimethylsilyloxy)silyl] silicate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only