Please login to your online account to display your discounted pricing

Alfa Aesar™ Octahydro-4(1H)-quinolinone, 97%

$287.80 - $287.80

Chemical Identifiers

Chemical Name or Material Octahydro-4(1H)-quinolinone
CAS 58869-89-9
Molecular Formula C9H15NO
Formula Weight 153.23
MDL Number MFCD02734410
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5018006

  SDS
alfa aesar™
H5018006
5g Each for $287.80
Description & Specifications

Specifications

Formula Weight 153.23
Chemical Name or Material Octahydro-4(1H)-quinolinone
Assay 97%
Molecular Formula C9H15NO
MDL Number MFCD02734410
CAS 58869-89-9
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Octahydro-4(1H)-quinolinone is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.