missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Octafluorotoluene, 98+%

Manufacturer:  Alfa Aesar™ A1145314

 View more versions of this product

Catalog No. AAA1145314



Specifications

Specifications

Octafluorotoluene
103°C to 105°C
25g
C7F8
UN1992
octafluorotoluene, perfluorotoluene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl benzene, toluene, octafluoro, benzene, pentafluoro trifluoromethyl, trifluoromethyl pentafluorobenzene, pentafluorobenzotrifluoride, 1,2,3,4,5-pentafluoro-6-trifluoromethyl-benzene, benzene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl, acmc-209juh
C1(=C(C(=C(C(=C1F)F)F)F)F)C(F)(F)F
236.064
236.06
20°C (68°F)
-66°C
434-64-0
MFCD00000375
2057072
USPWUOFNOTUBAD-UHFFFAOYSA-N
1,2,3,4,5-pentafluoro-6-(trifluoromethyl)benzene
9906
≥98%

Chemical Identifiers

434-64-0
236.064
USPWUOFNOTUBAD-UHFFFAOYSA-N
9906
C1(=C(C(=C(C(=C1F)F)F)F)F)C(F)(F)F
C7F8
MFCD00000375
octafluorotoluene, perfluorotoluene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl benzene, toluene, octafluoro, benzene, pentafluoro trifluoromethyl, trifluoromethyl pentafluorobenzene, pentafluorobenzotrifluoride, 1,2,3,4,5-pentafluoro-6-trifluoromethyl-benzene, benzene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl, acmc-209juh
1,2,3,4,5-pentafluoro-6-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H331-H315-H319-H335
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : XT3530000

TSCA : TSCA

SDS Documents