Learn More
Octafluoronaphthalene, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2169914
Description
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water. Solubility in toluene, almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Octafluoronaphthalene | |
84°C to 89°C | |
C10F8 | |
25 g | |
octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion | |
JDCMOHAFGDQQJX-UHFFFAOYSA-N | |
1,2,3,4,5,6,7,8-octafluoronaphthalene | |
67564 | |
96% |
313-72-4 | |
209°C | |
MFCD00014307 | |
1915630 | |
Insoluble in water. Solubility in toluene,almost transparent. | |
C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F | |
272.097 | |
272.1 |
Chemical Identifiers
313-72-4 | |
272.097 | |
JDCMOHAFGDQQJX-UHFFFAOYSA-N | |
67564 | |
C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F |
C10F8 | |
MFCD00014307 | |
octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion | |
1,2,3,4,5,6,7,8-octafluoronaphthalene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-239-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only