missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octafluorocyclopentene 98.0+%, TCI America™

Manufacturer:  TCI America O029250G

 View more versions of this product

Catalog No. O029250G



Specifications

Specifications

Octafluorocyclopentene
50g
C5F8
Perfluorocyclopentene
C1(=C(C(C(C1(F)F)(F)F)(F)F)F)F
212.042
212.04
≥98.0% (GC)
Colorless
559-40-0
MFCD00013745
YBMDPYAEZDJWNY-UHFFFAOYSA-N
1,2,3,3,4,4,5,5-octafluorocyclopentene
11212
Clear Liquid at 20°C

Chemical Identifiers

559-40-0
212.042
YBMDPYAEZDJWNY-UHFFFAOYSA-N
11212
C1(=C(C(C(C1(F)F)(F)F)(F)F)F)F
C5F8
MFCD00013745
Perfluorocyclopentene
1,2,3,3,4,4,5,5-octafluorocyclopentene
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4350

RTECSNumber : GY6000000

TSCA : Yes

Recommended Storage : Refrigerator

Documents