missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Octadecylsuccinic Anhydride 95.0+%, TCI America™
Supplier: TCI America O001825G
Specifications
Octadecylsuccinic Anhydride | |
47458-32-2 | |
25 g | |
C22H40O3 | |
octadecylsuccinic anhydride, n-octadecylsuccinic anhydride, octadecyl succinic anhydride, 2,5-furandione, dihydro-3-octadecyl, 3-octadecyldihydrofuran-2,5-dione, 2-octadecylsuccinic anhydride, dihydro-3-octadecylfuran-2,5-dione, stearylsuccinic anhydride, acmc-1an3y, 2, dihydro-3-octadecyl | |
CCCCCCCCCCCCCCCCCCC1CC(=O)OC1=O | |
352.559 | |
352.56 |
79°C | |
White-Yellow | |
Crystal-Powder at 20°C | |
MFCD00014551 | |
ZJFCVUTYZHUNSW-UHFFFAOYSA-N | |
3-octadecyloxolane-2,5-dione | |
96562 | |
≥95.0% (T) |
Chemical Identifiers
47458-32-2 | |
352.559 | |
ZJFCVUTYZHUNSW-UHFFFAOYSA-N | |
96562 | |
CCCCCCCCCCCCCCCCCCC1CC(=O)OC1=O |
C22H40O3 | |
MFCD00014551 | |
octadecylsuccinic anhydride, n-octadecylsuccinic anhydride, octadecyl succinic anhydride, 2,5-furandione, dihydro-3-octadecyl, 3-octadecyldihydrofuran-2,5-dione, 2-octadecylsuccinic anhydride, dihydro-3-octadecylfuran-2,5-dione, stearylsuccinic anhydride, acmc-1an3y, 2, dihydro-3-octadecyl | |
3-octadecyloxolane-2,5-dione |
Safety and Handling
EINECSNumber : (2)-0851&(2)-1134
TSCA : Yes