missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octadecylsuccinic Anhydride, TCI America™

$180.28 - $912.11

Chemical Identifiers

CAS 47458-32-2
Molecular Formula C22H40O3
Molecular Weight (g/mol) 352.559
MDL Number MFCD00014551
InChI Key ZJFCVUTYZHUNSW-UHFFFAOYSA-N
Synonym octadecylsuccinic anhydride, n-octadecylsuccinic anhydride, octadecyl succinic anhydride, 2,5-furandione, dihydro-3-octadecyl, 3-octadecyldihydrofuran-2,5-dione, 2-octadecylsuccinic anhydride, dihydro-3-octadecylfuran-2,5-dione, stearylsuccinic anhydride, acmc-1an3y, 2, dihydro-3-octadecyl
PubChem CID 96562
IUPAC Name 3-octadecyloxolane-2,5-dione
SMILES CCCCCCCCCCCCCCCCCCC1CC(=O)OC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O0018250G View Documents TCI America
O0018250G
250g Each for $912.11
 
O001825G View Documents TCI America
O001825G
25g Each for $180.28
 
Specifications

Chemical Identifiers

47458-32-2
352.559
ZJFCVUTYZHUNSW-UHFFFAOYSA-N
96562
CCCCCCCCCCCCCCCCCCC1CC(=O)OC1=O
C22H40O3
MFCD00014551
octadecylsuccinic anhydride, n-octadecylsuccinic anhydride, octadecyl succinic anhydride, 2,5-furandione, dihydro-3-octadecyl, 3-octadecyldihydrofuran-2,5-dione, 2-octadecylsuccinic anhydride, dihydro-3-octadecylfuran-2,5-dione, stearylsuccinic anhydride, acmc-1an3y, 2, dihydro-3-octadecyl
3-octadecyloxolane-2,5-dione

Specifications

White-Yellow
250g
C22H40O3
octadecylsuccinic anhydride, n-octadecylsuccinic anhydride, octadecyl succinic anhydride, 2,5-furandione, dihydro-3-octadecyl, 3-octadecyldihydrofuran-2,5-dione, 2-octadecylsuccinic anhydride, dihydro-3-octadecylfuran-2,5-dione, stearylsuccinic anhydride, acmc-1an3y, 2, dihydro-3-octadecyl
CCCCCCCCCCCCCCCCCCC1CC(=O)OC1=O
352.559
352.56
≥95.0% (T)
79°C
47458-32-2
MFCD00014551
ZJFCVUTYZHUNSW-UHFFFAOYSA-N
3-octadecyloxolane-2,5-dione
96562
Crystal-Powder at 20°C
Octadecylsuccinic Anhydride
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0851&(2)-1134

TSCA : Yes