Learn More
Octadecylmagnesium chloride, 0.3M solution in THF, AcroSeal™, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC436018000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Octadecylmagnesium chloride | |
6.96,87.82 | |
0.9060g/mL | |
67.0°C | |
C18H37ClMg | |
magnesium,chlorooctadecyl, n-octadecylmagnesium chloride, 0.25m w/w in thf, octadecylmagnesium chloride, octadecylmagnesium chloride solution, 0.5 m in thf, onjpxwzqvlkgiu-uhfffaoysa-m | |
Solubility in water: reacts. | |
0.906 | |
magnesium;octadecane;chloride | |
4192511 | |
Solution, Any Turbidity Can Occur |
116980-66-6,109-99-9 | |
12.18,93.04 | |
Brown to Yellow or Gray | |
−20°C | |
AcroSeal™ Glass Bottle | |
800mL | |
BNWJANSKVJZWMB-UHFFFAOYSA-M | |
CCCCCCCCCCCCCCCCC[CH2-].[Mg+2].[Cl-] | |
313.24 | |
313.24 |
Chemical Identifiers
116980-66-6 | |
313.24 | |
magnesium,chlorooctadecyl, n-octadecylmagnesium chloride, 0.25m w/w in thf, octadecylmagnesium chloride, octadecylmagnesium chloride solution, 0.5 m in thf, onjpxwzqvlkgiu-uhfffaoysa-m | |
magnesium;octadecane;chloride |
C18H37ClMg | |
BNWJANSKVJZWMB-UHFFFAOYSA-M | |
4192511 | |
CCCCCCCCCCCCCCCCC[CH2-].[Mg+2].[Cl-] |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.<br/
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
RUO – Research Use Only