Please login to your online account to display your discounted pricing

Octadecylmagnesium chloride, 0.3M solution in THF, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Octadecylmagnesium chloride
CAS 116980-66-6
CAS Min % 6.96
CAS Max % 12.18
CAS 109-99-9
CAS Min % 87.82
CAS Max % 93.04
Molecular Formula C18H37ClMg
Formula Weight 313.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC436018000

  SDS
thermo fisher scientific
AC436018000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC436011000

  SDS
thermo fisher scientific
AC436011000
AcroSeal™ Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solution, Any Turbidity Can Occur
Formula Weight 313.24
Quantity 800mL
Color Yellow to brown or gray
Packaging AcroSeal™ Glass Bottle
Density 0.9060g/mL
Chemical Name or Material Octadecylmagnesium chloride
Molecular Formula C18H37ClMg
CAS 116980-66-6
CAS Min % 6.96
Health Hazard 1 Danger
CAS Max % 12.18
CAS 109-99-9
CAS Min % 87.82
CAS Max % 93.04
Health Hazard 2 GHS P Statement Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Do not breathe dust/fume/gas/mist/vapors/spray. Wash face, hands and any exposed skin thoroughly after handling. IF IN EYES: R

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Do not breathe dust/fume/gas/mist/vapors/spray. Wash face, hands and any exposed skin thoroughly after handling. IF IN EYES: R

GHS P Statement Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides.