Please login to your online account to display your discounted pricing

Octadecylmagnesium chloride, 0.3M solution in THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC436011000

 View more versions of this product

Catalog No. AC436011000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Physical Form Solution, Any Turbidity Can Occur
Formula Weight 313.24
Quantity 100mL
Color Yellow to brown or gray
Packaging AcroSeal™ Glass Bottle
Density 0.9060g/mL
Chemical Name or Material Octadecylmagnesium chloride
CAS Min % 6.96
Health Hazard 1 Danger
Molecular Formula C18H37ClMg
CAS 116980-66-6
CAS 109-99-9
CAS Max % 93.04
CAS Max % 12.18
CAS Min % 87.82

Specifications

Chemical Name or Material Octadecylmagnesium chloride
CAS 116980-66-6
CAS Min % 6.96
CAS Max % 12.18
CAS 109-99-9
CAS Min % 87.82
CAS Max % 93.04
Molecular Formula C18H37ClMg
Formula Weight 313.24
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.

SDS & Safety Documents