missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octadecylamine Acetate, TCI America™

$36.67 - $155.84

Chemical Identifiers

CAS 2190-04-7
Molecular Formula C20H43NO2
Molecular Weight (g/mol) 329.57
MDL Number MFCD00060198
InChI Key UPHWVVKYDQHTCF-UHFFFAOYSA-N
Synonym Aminooctadecane Acetate, Stearylamine Acetate, Octadecan-1-amine Acetate
PubChem CID 255899
IUPAC Name acetic acid;octadecan-1-amine
SMILES CCCCCCCCCCCCCCCCCCN.CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O0015500G View Documents TCI America
O0015500G
500g Each for $155.84
 
O001525G View Documents TCI America
O001525G
25g Each for $36.67
 
Specifications

Chemical Identifiers

2190-04-7
329.57
UPHWVVKYDQHTCF-UHFFFAOYSA-N
255899
CCCCCCCCCCCCCCCCCCN.CC(=O)O
C20H43NO2
MFCD00060198
Aminooctadecane Acetate, Stearylamine Acetate, Octadecan-1-amine Acetate
acetic acid;octadecan-1-amine

Specifications

White-Yellow
500g
C20H43NO2
Aminooctadecane Acetate, Stearylamine Acetate, Octadecan-1-amine Acetate
CCCCCCCCCCCCCCCCCCN.CC(=O)O
329.57
329.57
≥60.0% (GC)
75°C
2190-04-7
MFCD00060198
UPHWVVKYDQHTCF-UHFFFAOYSA-N
acetic acid;octadecan-1-amine
255899
Crystal-Powder at 20°C
Octadecylamine Acetate
Safety and Handling

Safety and Handling

RTECSNumber : RG4170000

TSCA : Yes