missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Octadecylamine Acetate 60.0+%, TCI America™
Supplier: TCI America O001525G
Specifications
Octadecylamine Acetate | |
75°C | |
C20H43NO2 | |
25 g | |
UPHWVVKYDQHTCF-UHFFFAOYSA-N | |
acetic acid;octadecan-1-amine | |
255899 | |
≥60.0% (GC) |
2190-04-7 | |
White-Yellow | |
MFCD00060198 | |
Aminooctadecane Acetate, Stearylamine Acetate, Octadecan-1-amine Acetate | |
CCCCCCCCCCCCCCCCCCN.CC(=O)O | |
329.57 | |
329.57 | |
Crystalline Powder |
Chemical Identifiers
2190-04-7 | |
329.57 | |
UPHWVVKYDQHTCF-UHFFFAOYSA-N | |
255899 | |
CCCCCCCCCCCCCCCCCCN.CC(=O)O |
C20H43NO2 | |
MFCD00060198 | |
Aminooctadecane Acetate, Stearylamine Acetate, Octadecan-1-amine Acetate | |
acetic acid;octadecan-1-amine |
Safety and Handling
RTECSNumber : RG4170000
TSCA : Yes