missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octadecyl 3-Mercaptopropionate (contains ca. 12% Hexadecyl 3-Mercaptopropionate) 85.0+%, TCI America™

Manufacturer:  TCI America O036225G

 View more versions of this product

Catalog No. O036225G



Specifications

Specifications

Octadecyl 3-Mercaptopropionate (contains ca. 12% Hexadecyl 3-Mercaptopropionate)
25g
C21H42O2S
3-Mercaptopropionic Acid Octadecyl Ester, Stearyl 3-Mercaptopropionate, 3-Mercaptopropionic Acid Stearyl Ester
CCCCCCCCCCCCCCCCCCOC(=O)CCS
358.625
358.63
≥85.0% (GC)
White
31778-15-1
MFCD00046844
UCUPUEARJPTGKU-UHFFFAOYSA-N
octadecyl 3-sulfanylpropanoate
169361
Crystal-Lump at 20°C

Chemical Identifiers

31778-15-1
358.625
UCUPUEARJPTGKU-UHFFFAOYSA-N
169361
CCCCCCCCCCCCCCCCCCOC(=O)CCS
C21H42O2S
MFCD00046844
3-Mercaptopropionic Acid Octadecyl Ester, Stearyl 3-Mercaptopropionate, 3-Mercaptopropionic Acid Stearyl Ester
octadecyl 3-sulfanylpropanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1383

TSCA : Yes

Documents