missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Octadecenylsuccinic Anhydride (mixture of isomers) 93.0+%, TCI America™
Supplier: TCI America O0013500G
Specifications
Octadecenylsuccinic Anhydride (mixture of isomers) | |
28777-98-2 | |
500 g | |
C22H38O3 | |
3-octadec-1-en-1-yl dihydrofuran-2,5-dione, 3-1e-octadec-1-en-1-yl oxolane-2,5-dione, n-octadecenylsuccinic anhydride, dsstox_cid_7948, dsstox_rid_78620, dsstox_gsid_27948, n-octadecenyl succinic anhydride, 3-e-octadec-1-enyl oxolane-2,5-dione, 2,5-furandione, dihydro-3-1-octadecen-1-yl, n-octadecenylsuccinic anhydride, mixture of isomers | |
CCCCCCCCCCCCCCCCC=CC1CC(=O)OC1=O | |
350.543 | |
350.54 |
70°C | |
White-Yellow | |
Crystal-Powder at 20°C | |
MFCD00014545 | |
KAYAKFYASWYOEB-ISLYRVAYSA-N | |
3-[(E)-octadec-1-enyl]oxolane-2,5-dione | |
5354626 | |
≥93.0% (GC) |
Chemical Identifiers
28777-98-2 | |
350.543 | |
KAYAKFYASWYOEB-ISLYRVAYSA-N | |
5354626 | |
CCCCCCCCCCCCCCCCC=CC1CC(=O)OC1=O |
C22H38O3 | |
MFCD00014545 | |
3-octadec-1-en-1-yl dihydrofuran-2,5-dione, 3-1e-octadec-1-en-1-yl oxolane-2,5-dione, n-octadecenylsuccinic anhydride, dsstox_cid_7948, dsstox_rid_78620, dsstox_gsid_27948, n-octadecenyl succinic anhydride, 3-e-octadec-1-enyl oxolane-2,5-dione, 2,5-furandione, dihydro-3-1-octadecen-1-yl, n-octadecenylsuccinic anhydride, mixture of isomers | |
3-[(E)-octadec-1-enyl]oxolane-2,5-dione |
Safety and Handling
EINECSNumber : (2)-0852
RTECSNumber : LU1245000
TSCA : Yes