missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octadecane 98.0+%, TCI America™

Manufacturer:  TCI America O0003250G

 View more versions of this product

Catalog No. O0003250G



Specifications

Specifications

Octadecane
250g
C18H38
n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group
CCCCCCCCCCCCCCCCCC
254.502
CHEBI:32926
Crystal-Lump at 20°C
White
593-45-3
MFCD00009007
RZJRJXONCZWCBN-UHFFFAOYSA-N
octadecane
11635
254.50
≥98.0% (GC)

Chemical Identifiers

593-45-3
254.502
RZJRJXONCZWCBN-UHFFFAOYSA-N
11635
octadecane
C18H38
MFCD00009007
n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group
CHEBI:32926
CCCCCCCCCCCCCCCCCC
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0010

TSCA : Yes

Documents