Please login to your online account to display your discounted pricing

Octadecanamide (Tech.), ca 60% C18, ACROS Organics™


Quantity 1kg

Manufacturer: thermo fisher scientific  129280010

 View more versions of this product

Catalog No. AC129280010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥55%
Physical Form Granules
Purity Grade Technical Grade
Formula Weight 283.5
Quantity 1kg
Color Light yellow
TSCA TSCA
Melting Point 96° to 104°C
RTECS Number RG0182000
EINECS Number 204-693-2
Beilstein 02, 384
Acid Value 9mg KOH/g max.
Iodine Number 3g I2/100g max.
Water 0.15% max. (K.F.)
Synonym Stearamide
Infrared Spectrum Authentic
Chemical Name or Material Octadecanamide
CAS 124-26-5
Linear Formula CH3(CH2)16CONH2
Name Note tech., ca 60% C18
Molecular Formula C18H37NO

Specifications

Chemical Name or Material Octadecanamide
Name Note tech., ca 60% C18
CAS 124-26-5
Purity Grade Technical Grade
Assay ≥55%
Molecular Formula C18H37NO
Linear Formula CH3(CH2)16CONH2
Formula Weight 283.5
Synonym Stearamide