Please login to your online account to display your discounted pricing

Octadecanamide (Tech.), ca 60% C18, ACROS Organics™

$168.66 - $168.66

Chemical Identifiers

Chemical Name or Material Octadecanamide
Name Note tech., ca 60% C18
CAS 124-26-5
Purity Grade Technical Grade
Assay ≥55%
Molecular Formula C18H37NO
Linear Formula CH3(CH2)16CONH2
Formula Weight 283.5
Synonym Stearamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC129280010

  SDS
thermo fisher scientific
129280010
1kg Each for $168.66
Description & Specifications

Specifications

Assay ≥55%
Physical Form Granules
Purity Grade Technical Grade
Formula Weight 283.5
Quantity 1kg
Color Light yellow
TSCA TSCA
Melting Point 96° to 104°C
RTECS Number RG0182000
EINECS Number 204-693-2
Beilstein 02, 384
Acid Value 9mg KOH/g max.
Iodine Number 3g I2/100g max.
Water 0.15% max. (K.F.)
Synonym Stearamide
Infrared Spectrum Authentic
Chemical Name or Material Octadecanamide
CAS 124-26-5
Linear Formula CH3(CH2)16CONH2
Name Note tech., ca 60% C18
Molecular Formula C18H37NO