Please login to your online account to display your discounted pricing

Alfa Aesar™ Octadecafluorodecahydronaphthalene, 95+%

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1581606

 
alfa aesar™
15816-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1581614

 
alfa aesar™
15816-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 1.908g/mL
Refractive Index 1.3145
Quantity 5g
Boiling Point 142°C
Melting Point −10°C
Quantity 5g
Flash Point 40° (105°F)
Odor Odorless
Chemical Name or Material Octadecafluorodecahydronaphthalene
CAS 306-94-5
Physical Form Liquid
Assay ≥95%
Formula Weight 462.07g/mol
UN Number UN1993
Merck Index 14,4183
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.