missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Octacosane, MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 442696
Description
PetroleumSpecifications
Ampule | |
630-02-4 | |
CH3(CH2)26 CH3 | |
394.76 | |
<1 mmHg (20°C) | |
57°C to 64°C | |
Neat |
1000 mg | |
Analytical Standard | |
MFCD00009355 | |
Analytical Standard | |
13.6 (vs. air) | |
278°C (15 mmHg) |