missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Octacosane, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 129270050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Octacosane | |
99.0 | |
61.0°C to 64.0°C | |
0.8067g/mL | |
227°C | |
99% min. (GC) | |
C28H58 | |
MFCD00009355 | |
01,I,71 | |
n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g | |
ZYURHZPYMFLWSH-UHFFFAOYSA-N | |
octacosane | |
12408 | |
394.76 | |
Shiny Flakes or Powder |
630-02-4 | |
100.0 | |
White | |
440.0°C | |
Authentic | |
Glass bottle | |
CH3(CH2)26CH3 | |
5 g | |
0.8067 | |
Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
394.77 | |
CHEBI:32943 | |
99+% |
Chemical Identifiers
630-02-4 | |
394.77 | |
ZYURHZPYMFLWSH-UHFFFAOYSA-N | |
12408 | |
octacosane |
C28H58 | |
MFCD00009355 | |
n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g | |
CHEBI:32943 | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Safety and Handling
EINECSNumber : 211-125-7
RUO – Research Use Only