missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OCT3/4 Rat anti-Human, Mouse, eFluor 570, Clone: EM92, eBioscience™

Rat Monoclonal Antibody

$174.24 - $456.75

Specifications

Antigen OCT3/4
Clone EM92
Host Species Rat
Gene Alias OCT3, OCT4, octamer-binding transcription factor, POU5F1
Applications Immunocytochemistry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123635 Life Technologies
LS41584180
25μg Each for $174.24
501123636 Life Technologies
LS41584182
100μg Each for $456.75
Specifications

Description

The EM92 monoclonal antibody reacts with mouse and human Oct3/4, encoded by the Pou5F1 gene. Oct3/4 is a POU domain-containing transcription factor that is critical for maintaining embryonic stem (ES) and induced pluripotent stem (iPS) cells in a pluripotent state, and is expressed by ES, embryonic germ cells and embryonic carcinoma cell lines. In cells of the inner cell mass (ICM), reduction of Oct3/4 expression causes dedifferentiation to trophoectoderm, whereas increased expression results in differentiation to mesoderm and primitive endoderm. Oct3/4 regulates the expression of several genes, including FGF-4, UTF1, Sox2, Fbx15, Rex1 and osteopontin through distinct mechanisms. Furthermore, Oct3/4 frequently acts synergistically with Sox2 to regulate target gene expression, as is the case with FGF-4. It has been demonstrated that Oct3/4 expression in ES cells can be negatively regulated by either treatment with retinoic acid, or by removal of leukemia-inhibitory factor (LIF).
Description & Specifications

Specifications

OCT3/4
Rat
Immunocytochemistry
eFluor 570
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
EM92
OCT3, OCT4, octamer-binding transcription factor, POU5F1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer