Please login to your online account to display your discounted pricing

OCT3/4 Rat anti-Human, Mouse, Clone: EM92, eBioscience™

Rat Monoclonal Antibody

$201.55 - $404.55

Specifications

Antigen OCT3/4
Clone EM92
Host Species Rat
Gene Alias OCT3, OCT4, octamer-binding transcription factor, POU5F1
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013203

 
life technologies
LS14584180
25μg Each for $201.55

5013204

 
life technologies
LS14584182
100μg Each for $404.55
Description & Specifications

Specifications

Antigen OCT3/4
Clone EM92
Host Species Rat
Gene Alias OCT3, OCT4, octamer-binding transcription factor, POU5F1
Species Reactivity Human, Mouse
Applications Microscopy
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The EM92 monoclonal antibody reacts with mouse and human Oct3/4, encoded by the Pou5F1 gene. Oct3/4 is a POU domain-containing transcription factor that is critical for maintaining embryonic stem (ES) and induced pluripotent stem (iPS) cells in a pluripotent state, and is expressed by ES, embryonic germ cells and embryonic carcinoma cell lines. In cells of the inner cell mass (ICM), reduction of Oct3/4 expression causes dedifferentiation to trophoectoderm, whereas increased expression results in differentiation to mesoderm and primitive endoderm. Oct3/4 regulates the expression of several genes, including FGF-4, UTF1, Sox2, Fbx15, Rex1 and osteopontin through distinct mechanisms. Furthermore, Oct3/4 frequently acts synergistically with Sox2 to regulate target gene expression, as is the case with FGF-4. It has been demonstrated that Oct3/4 expression in ES cells can be negatively regulated by either treatment with retinoic acid, or by removal of leukemia-inhibitory factor (LIF).