missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Oct-2, Rabbit Polyclonal Antibody

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Oct-2, Rabbit Polyclonal Antibody.

$289.51 - $1,156.47

Specifications

Antigen 2-Oct
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human,Mouse,Human,Mouse
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RB9297P Thermo Scientific™ Lab Vision™
RB9297P
1mL Each for $1,156.47
RB9297P0 Thermo Scientific™ Lab Vision™
RB9297P0
100μL Each for $289.51
RB9297P1 Thermo Scientific™ Lab Vision™
RB9297P1
500μL Each for $699.51
RB9297R7 Thermo Scientific™ Lab Vision™
RB9297R7
7mL Each for $550.06
Specifications

Description

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Oct-2, Rabbit Polyclonal Antibody.

Oct-2 is a transcription factor belonging to the POU homeodomain family that binds to the Ig gene octomer sites regulating B cell specific genes. Oct-2 protein expression is not restricted to B cells, although expression levels are much higher in these cells. Oct-2 expression is reported to be significantly greater in germinal center derived lymphomas, although other B cell lymphomas also display high levels of expression.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide derived from C-terminus of human Oct-2

Molecular Weight: ∽80kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

2-Oct
Polyclonal
Human,Mouse,Human,Mouse
RUO
Primary
IgG
Immunohistochemistry (Paraffin)
Unlabeled
Rabbit
Transcription Regulation
Synthetic peptide derived from C-terminus of human Oct-2