Please login to your online account to display your discounted pricing

Ocinaplon, Tocris™

GABAA modulator; anxiolytic

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material Ocinaplon
Assay Percent Range >98%
Molecular Formula C17H11N5O
Formula Weight 301.3
Synonym 2-Pyridinyl[7-(4-pyridinyl)pyrazolo[1, 5-a]pyrimidin-3-yl]methanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

441010

 
r & d systems
441010
10mg Each for $239.63

441050

 
r & d systems
441050
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 301.3
Quantity 10mg
Chemical Name or Material Ocinaplon
Molecular Formula C17H11N5O
Recommended Storage Store at +4°C
CAS 96604-21-6
Synonym 2-Pyridinyl[7-(4-pyridinyl)pyrazolo[1, 5-a]pyrimidin-3-yl]methanone
Assay Percent Range >98%
Safety and Handling

Store at +4°C