Medicom™ Ocean Pacific™ Lab Master™ Nitrile Medical Examination Gloves with Textured Fingertips
Ocean Pacific™ Lab Master™ Nitrile Medical Examination Gloves with Textured Fingertips. Provides the protection and strength of nitrile without the chemical accelerators commonly found in nitrile gloves. Textured fingertips for a secure grip, even when wet. Chemo-tested.
Supplier:  Medicom™ MEDOPSLM2
 View more versions of this product
Catalog No. 296366821
Description
Description
  • These versatile all-purpose gloves provide all the protection and strength of nitrile without the chemical accelerators commonly found in nitrile gloves.
  • The DermaSense technology eliminates accelerators to reduce the risk of skin reactions.
  • Chemo-tested, accelerator-free, safe for food handling.
  • Designed for wider palm, shorter fingers.
Specifications
Specifications
No
Healthcare, Industrial, Laboratory
Indigo
Fingertip Textured
Powder Free
Non-sterile
Beaded
Yes
1.5
Single-use
Yes
Small
Yes