Please login to your online account to display your discounted pricing

American Educational Products Ocean Floor Raised-Relief Map


Dramatic map of the world captures the geography of the land and the ocean floor.

Manufacturer: american educational products  436

 View more versions of this product

Catalog No. S85207

  • / Each
 

Description & Specifications

Specifications

Product Type Raised Relief Map of the Ocean Floor
For Use With (Application) Dramatic map of the world captures the geography of the land and the ocean floor
Grade 3 to 12
Includes Making an ocean floor profile, studying continental margin types, earthquake and volcano distribution
Quantity Each