Please login to your online account to display your discounted pricing

American Educational Products Ocean Floor Raised-Relief Map

Dramatic map of the world captures the geography of the land and the ocean floor.

$67.50 - $101.25

Specifications

For Use With (Application) Dramatic map of the world captures the geography of the land and the ocean floor
Grade 3 to 12
Includes Making an ocean floor profile, studying continental margin types, earthquake and volcano distribution
Quantity Each
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S85207A

 
american educational products
446
Activity Guide Each for $101.25

S85207

 
american educational products
436
Raised Relief Map of the Ocean Floor Each for $67.50
Description & Specifications

Specifications

For Use With (Application) Dramatic map of the world captures the geography of the land and the ocean floor
Grade 3 to 12
Includes Making an ocean floor profile, studying continental margin types, earthquake and volcano distribution
Quantity Each

  • Depicts ridges, trenches, mountains, continental shelves, continental slopes, and islands
  • Unframed, 26 × 39 in. (66 × 99cm)
Teacher's Guide activities include:
  • Making an ocean floor profile
  • Studying continental margin types
  • Earthquake and volcano distribution