Thermo Scientific™ Objective Lenses for ArrayScan™ and CellInsight™ High-Content Platforms DFS Item
Expand the functionality of your Thermo Scientific™ high-content instrument
Manufacturer:  Thermo Scientific™ N010098
 View more versions of this product
Catalog No. N010098
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
Description & Specifications
Specifications
ArrayScan™ HCA Readers
Zeiss 20X Objective, Plan-Apochromat (0.8 NA)