Please login to your online account to display your discounted pricing

OBAA, Tocris


Phospholipase A2 inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS060610

 View more versions of this product

Catalog No. 060610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 428.65
Assay Percent Range >98%
Synonym 4-(4-Octadecylphenyl)-4-oxobutenoic acid
Recommended Storage Desiccate at +4°C
Chemical Name or Material OBAA
CAS 221632-26-4
Molecular Formula C28H44O3

Specifications

Chemical Name or Material OBAA
Assay Percent Range >98%
Molecular Formula C28H44O3
Formula Weight 428.65
Synonym 4-(4-Octadecylphenyl)-4-oxobutenoic acid
Accessories

Desiccate at +4°C