Please login to your online account to display your discounted pricing

OBAA, Tocris

Phospholipase A2 inhibitor

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material OBAA
Assay Percent Range >98%
Molecular Formula C28H44O3
Formula Weight 428.65
Synonym 4-(4-Octadecylphenyl)-4-oxobutenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

060650

 
r & d systems
RDS060650
50mg Each for $682.67

060610

 
r & d systems
RDS060610
10mg Each for $164.61
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 428.65
Chemical Name or Material OBAA
Assay Percent Range >98%
Molecular Formula C28H44O3
Synonym 4-(4-Octadecylphenyl)-4-oxobutenoic acid
Recommended Storage Desiccate at +4°C
CAS 221632-26-4
Safety and Handling

Desiccate at +4°C