missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OBAA, Tocris

Phospholipase A2 inhibitor

Manufacturer:  R & D Systems RDS060650

 View more versions of this product

Catalog No. 060650


 

Specifications Description & Specifications

Specifications

OBAA
>98%
428.65
50mg
221632-26-4
C28H44O3
4-(4-Octadecylphenyl)-4-oxobutenoic acid

Chemical Identifiers

>98%
428.65
OBAA
C28H44O3
4-(4-Octadecylphenyl)-4-oxobutenoic acid

Safety and Handling

Recommended Storage : Desiccate at +4°C