Please login to your online account to display your discounted pricing
Oakton™ Waterproof pHTestr BNC Electrodes and Accessories
Manufacturer: oakton™  3562470
 View more versions of this product
Catalog No. 13-300-208
  • / Each
Description & Specifications
Specifications
Connector Types BNC
For Use With Oakton pHTestr
Type Calibration kit
Includes Calibration buffer pouches (two each of pH 4.01, 7, 10, rinse water), sample jar, hard carrying case