missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oakton™ Waterproof pH 450 Portable Meter

Rugged design with backlit display for use in any environment

Manufacturer:  Oakton™ 3561890

 View more versions of this product

Catalog No. 15-500-122



Description

Description

Rugged design with backlit display for use in any environment

  • Intuitive, user-friendly icons show electrode status as pH slope
  • IP67 waterproof
  • Grip-Clip™ holder for quick and easy attachment of electrode to beaker or container
  • Large, backlit LCD displays measurement simultaneously with temperature and calibration data with % slope
  • Up to 6-point calibration from USA, NIST™ DIN and custom pH buffers
  • Automatic temperature compensation (ATC) for measurement accuracy, even in fluctuating temperatures
  • Perform data logging with manual or timed collection of up to 500 data sets with real-time clock time/date stamp
  • Download and analyze data sets with USB or RS-232 output
  • Measuring Range: -2.00 to 16.00 (pH); ±2000 (mV); -10° to +110°C (temp.); 0.0 to 19,999 (Ion)

  • Select temperature readout from °C/°F to suit your application
  • Other features include stability indicator, smart averaging, hold function, visible calibration alarm, battery life indicator, electrode status
  • Two AA batteries with extended battery life or optional universal power adapter
  • Built-in stand with wall mount option allows for easy benchtop measurement or storage

pH 450 Portable Meter Models

  • Cat. No. 15-500-119: Includes Grip-Clip probe holder and two AA batteries
  • Cat. No. 15-500-121: Includes NIST-Traceable calibration, Grip-Clip probe holder and two AA batteries
  • Cat. No. 15-500-118: Includes All-in-one pH/temperature electrode, Grip-Clip probe holder and two AA batteries
  • Cat. No. 15-500-120: Includes NIST™-Traceable calibration, All-in-one pH/temperature electrode, Grip-Clip probe holder and two AA batteries
  • Cat. No. 15-500-122: Includes All-in-one pH/temperature electrode, Grip-Clip probe holder, buffers and electrode storage solution, two AA batteries and hard carrying case
  • Cat. No. 15-500-123: Includes NIST-Traceable calibration, All-in-one pH/temperature electrode, Grip-Clip probe holder, buffers and electrode storage solution, two AA batteries and hard carrying case

Applications:

Ideal for manufacturing floor, field testing and laboratory

Certifications

CE marked

Warranty

Three years

Specifications

Specifications

500 Data Sets
Automatic or Manual
17° to 230°F
pH Meter Kit
Optional universal AC adapter
±0.5% full-scale (monovalent); ±1% full-scale (divalent)
±0.2
500 hr.
pH: Up to 6-points (USA, NIST, DIN); ±200mV; Up to 6 points (Ion); 0.1 increments (temp.)
CE
5.7cm
0.01
2.25 in.
IP-67
20cm
All-in-one pH/temperature electrode
0.01
±2000 mV
LCD (Graphic with Backlight)
0.1°
-10° to +110°C
3.12 in.
±0.01
8cm
±0.5°
2 x AA
Manual or timed collection
20 x 8 x 5.7cm (7.87 x 3.12 x 2.25 in.)
Manufacturing floor, field testing and laboratory
-2.00 to 16.00
All-in-one pH/temperature electrode, Grip-Clip electrode holder, buffers and electrode storage solution, two AA batteries, hard carrying case
7.87 in.
USB, RS-232
0.0 to 19,999
0.1mV ±999.9mV; 1mV beyond
Documents