missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oakton™ Waterproof pH 150 Portable Meter

Rugged, reliable and accurate meter to measure pH, mV and temperature

Manufacturer:  Oakton™ 3561433

 View more versions of this product

Catalog No. 15-500-114



Description

Description

Rugged, reliable and accurate meter to measure pH, mV and temperature

  • Intuitive, user-friendly icons show electrode status as pH slope
  • IP67 waterproof
  • Grip-Clip™ holder for quick and easy attachment of electrode to beaker or container
  • Large, three-line LCD displays pH or mV measurement simultaneously with temperature and calibration data with % slope
  • Up to 6-point calibration from USA, NIST™ and DIN pH buffer sets
  • Auto buffer recognition feature to identify correct pH buffer for rapid calibration
  • Automatic temperature compensation (ATC) for measurement accuracy, even in fluctuating temperatures
  • mV offset for calibration of ORP standards, zeroing mV value, and pH diagnostics
  • Measuring Range: -2.00 to 16.00 (pH); ±2000 (mV); -10° to +110°C (temp.)

  • Manual data storage allows recall of 150 data sets, as well as the last calibration's data
  • Select temperature readout from °C/°F to suit your application
  • Other features include stability indicator, smart averaging, hold function, visible calibration alarm, battery life indicator, BNC connector and electrode status
  • Two AA batteries with 700 hours battery-life or optional universal power adapter
  • Built-in stand with wall mount option allows for easy benchtop measurement or storage

pH 150 Portable Meter Models

  • Cat. No. 15-500-113: Includes Grip-Clip electrode holder and two AA batteries
  • Cat. No. 15-500-115: Includes NIST-Traceable calibration provided by InnoCal, Grip-Clip electrode holder and two AA batteries
  • Cat. No. 15-500-112: Includes All-in-One single-junction pH/ATC electrode, Grip-Clip electrode holder and two AA batteries
  • Cat. No. 15-500-114: Includes NIST™-Traceable calibration provided by InnoCal, All-in-One single-junction pH/ATC electrode, Grip-Clip electrode holder and two AA batteries
  • Cat. No. 15-500-116: Includes All-in-One single-juntion pH/ATC electrode, Grip-Clip electrode holder, calibration solutions, carrying case and two AA batteries
  • Cat. No. 15-500-117: Includes NIST-Traceable calibration certificate provided by InnoCal, All-in-One single-junction pH/ATC electrode, Grip-Clip electrode holder, calibration solutions, carrying case and two AA batteries

Certifications

CE marked

Warranty

Three years

Specifications

Specifications

Automatic or Manual
17° to 230°F
pH Meter with Electrode and NIST-Traceable calibration
Optional universal AC adapter
8cm
±0.5°
2 x AA
Manual storage
20 x 8 x 5.7cm (7.87 x 3.12 x 2.25 in.)
LCD
Measuring pH, mV and temperature
-2.00 to 16.00
NIST-Traceable calibration provided by InnoCal, All-in-One single-junction pH/ATC electrode, Grip-Clip electrode holder and two AA batteries
7.87 in.
All-in-One single-junction pH/ATC electrode
±2000mV
0.1°
-10° to +110°C
3.12 in.
±0.01
±0.2
700 hr.
pH: Up to 6-points (USA, NIST, DIN); ±200mV; 0.1° increments (temp.)
CE
150 data sets
5.7cm
0.01
2.25 in.
IP-67
20cm
0.1 from 0.1 to 999.9, 1 from 1000 to 2000
Documents