missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oakton™ Waterproof PC 450 Portable Multiparameter Meter

Choose meter with combination pH/conductivity/temperature probe or separate pH and conductivity/temperature probes

$1,929.60 - $2,477.20

Specifications

For Use With (Application) Ideal for manufacturing floor, field testing, as well as the laboratory.
Range (Conductivity) 0 to 200mS
Accuracy (Conductivity) ±1% full-scale
Resolution (Conductivity) 0.01μS to 0.1mS
Range (mV) ±2000
View More Specs

Products 4
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
15-500-136 View Documents Oakton™
3563010
Multiparameter Meter with Combination Probe Each for $1,929.60
 
15-500-137 View Documents Oakton™
3563011
Multiparameter Meter with combination Probe and calibration Each for $2,388.33
 
15-500-138 View Documents Oakton™
3563012
Multiparameter Meter with separate pH and conductivity/ATC probes Each for $2,006.84
 
15-500-139 View Documents Oakton™
3563013
Multiparameter Meter with separate pH and conductivity/ATC probes and Calibration Each for $2,477.20
 
Description

Description

  • Rugged, IP67 waterproof housing
  • Intuitive, user-friendly icons; electrode status displays pH slope
  • Large, backlit custom LCD to view main measurement simultaneously with temperature
  • Grip-Clip™ electrode holder quickly and easily secures electrode to side of beaker or container (for use with separate pH and conductivity/ATC probes)
  • Calibrate pH with up to six points from USA, NIST, DIN, and custom pH buffer sets
  • Autocal feature for conductivity has one calibration point per range
  • ATC ensures measurement accuracy even in fluctuating temperatures
  • Memory of 500 data sets and USB or RS-232 output for transfer to PC
  • Other features include: ready (stability) indicator, smart averaging, user-selectable °C/°F units, hold function, calibration alarm, battery life-indicator
  • Perform data logging with manual or timed collection of up to 500 data sets with real-time clock time/date stamp
  • Extended battery life or optional universal power adapter
  • Built-in stand with wall mount allows for easy benchtop measurement or storage

Specifications:
  • pH Range: -2.00 to 16.0
  • pH Resolution: 0.01
  • pH Accuracy: ±0.01
  • mV Range: ±2000
  • mV Resolution: 0.1 to 1
  • mV Accuracy: ±0.5
  • Conductivity Range: 0 to 200mS
  • Conductivity Resolution: 0.01μS to 0.1mS
  • Conductivity Accuracy: ±1% full-scale
  • TDS Range: 0 to 200 ppt
  • TDS Resolution: 0.01ppm to 0.1 ppt
  • TDS Accuracy: ±1% full-scale
  • Salinity Range: 0 to 50 ppt
  • Salinity Resolution: 0.1 ppt
  • Salinity Accuracy: ±1% full-scale
  • Temperature Range: -10° to +110°C (17° to 230°F)
  • Temperature Resolution: 0.1
  • Temperature Accuracy: ±0.5°C (±0.9°F)
  • Temperature Compensation: Manual/Automatic
  • Interface: USB, RS232
  • Power: Two AA batteries, optional AC adapter
  • Battery Life: 500 hours
  • Dimensions (L x W x H): 20 x 8 x 5.7cm (7.9 x 3.12 x 2.25 in.)
Applications:
Ideal for manufacturing floor, field testing, as well as the laboratory.


Includes:
  • Cat. No. 15-500-136: Meter, combination pH/conductivity/temperature probe and two AA batteries
  • Cat. No. 15-500-137: Meter, combination pH/conductivity/temperature probe, two AA batteries and NIST-Traceable calibration certificate provided by InnoCal
  • Cat. No. 15-500-138: Meter, pH probe, conductivity/temperature cell K=1.0, Grip-Clip holder and two AA batteries
  • Cat. No. 15-500-139: Meter, pH probe, conductivity/temperature cell K=1.0, Grip-Clip holder, two AA batteries and NIST-Traceable calibration certificate provided by InnoCal

Order Info

Order optional data cable for USB or RS-232 connectivity to transfer data to a PC separately.

Certifications

CE marked, TUV 3-1, FCC Class A

Warranty

Three years

Specifications

Specifications

Ideal for manufacturing floor, field testing, as well as the laboratory.
±1% full-scale
±2000
0.1 to 1
±0.01
0 to 50 ppt
0.1 ppt
±1% full-scale
-10° to +110°C
±0.5°C (±0.9°F)
IP67
Three years
One calibration point per range
Two AA batteries
20cm
3.12 in.
2.25 in.
0 to 200mS
0.01μS to 0.1mS
±0.5
-2.00 to 16.0
0.01
±1% full-scale
0 to 200 ppt
0.01ppm to 0.1 ppt
17° to 230°F
Backlit LCD
CE marked, TUV 3-1, FCC Class A
Up to to six points from USA, NIST, DIN, and custom pH buffer sets
150 sets
7.9 in.
8cm
5.7cm