Please login to your online account to display your discounted pricing

Oakton™ Waterproof Big Display pHTestr™ 30


Dual Display of pH and Temperature plus high accuracy

Manufacturer: oakton™  3563430

 View more versions of this product

Catalog No. 13-200-263

  • / Each
 

Description & Specifications

Specifications

Includes Meter, electrode, four 1.5V batteries, instructions
Range (pH) -1 to 15
Resolution (pH) 0.01
Accuracy (pH) ±0.01
Calibration Points (pH) Up to 3
Resolution (Temperature) 0.1°C
Accuracy (Temperature) ±1°C
Display Type LCD
IP Rating 67
Certifications/Compliance USA, NIST
Temperature (English) Operating 32° to 122°F
Temperature (Metric) Operating 0 to 50°C
Detection Range -1 to +15pH
Diameter (English) 1.5 in.
Diameter (Metric) 3.8cm
For Use With (Application) Hold function, auto-off, error messages, waterproof, dustproof and floatable
Resolution 0.1pH
Shipping Weight (English) 4.5 oz.
Shipping Weight (Metric) 0.12kg
Accuracy ±0.1pH
Length (Metric) 16.5cm
Warranty 1 Year
Accessories

Accessories for Catalog Number: 13200263

Item Mfr. No. Item Description Price Per Unit Quantity  
13-300-149
Oakton™ buffer pouches, Cole-Parmer Instrument Company
oakton™
3565302
OAKTON pH buffer pouches; pH 7.00 package of 20 Economical pouches are accurate and convenient.±0.01 accuracy at 77°°F (25°°C) Pack of 20 Each for $89.34
13-300-148
Oakton™ pH buffer pouches
oakton™
WD3565303
OAKTON pH buffer pouches; package of 20 Economical pouches are accurate and convenient.±0.01 accuracy at 77°°F (25°°C) Pack of 20 Each for $89.34
SB105
Buffer-Pac Color-Coded Solutions (Certified), Fisher Chemical
fisher scientific company
BPSB105
Pack contains 1 each: SB101-500, pH 4.00 (Red), SB107-500, pH 7.00 (Yellow), SB115-500, pH 10.00 (Blue) Pack of 3 Each for $182.23
Case of 6 Pack for $1,093.38
13-300-147
Fisher Scientific™ Assorted pH Buffer Pouches
fisher scientific™
WD3565304
Economical, accurate, convenient Each for $86.86
S65290
Replacement Sensor for Oakton™ Double-Junction Waterproof pHTestr™
oakton instruments
3562438
Replaceable sensor saves money by reusing the same meter body Each for $210.66
13-300-150
Oakton™ pH buffer pouches
oakton™
WD3565301
OAKTON pH buffer pouches; package of 20 Economical pouches are accurate and convenient.±0.01 accuracy at 77°°F (25°°C) Pack of 20 Each for $89.34