Please login to your online account to display your discounted pricing

Oakton™ Waterproof Big Display phTestr™ 10

Pocket pH meter with large display makes taking readings easy

$194.47 - $261.60

Specifications

Accuracy (pH) ±0.1
Calibration Points (pH) Up to 3
Certifications/Compliance USA, NIST
Diameter (English) 1.5 in.
Diameter (Metric) 3.8cm
View More Specs
Includes:  Meter Only package includes meter, electrode, four 1.5V batteries, instructions. Meter Kit package includes meter; electrode; two each of pH 4.01, 7.00, 10.00 buffers and rinse solution in single-use pouches, sample jar, batteries, instructions and hard Plastic carrying case
Products
Catalog Number Mfr. No. Includes Price Quantity    

13-200-268

 
oakton™
WD3563470
Meter, electrode, two each of pH 4.01, 7, 10 Buffers and rinse solution in single-use pouches, sample jar, batteries, instructions and hard Plastic carrying case Each for $261.60

13-200-261

 
oakton™
3563410
Meter, electrode, four 1.5V batteries, instructions Each for $194.47
Description & Specifications

Specifications

Accuracy (pH) ±0.1
Calibration Points (pH) Up to 3
Certifications/Compliance USA, NIST
Diameter (English) 1.5 in.
Diameter (Metric) 3.8cm
Display Type LCD
IP Rating 67
Range (pH) -1 to 15
Resolution (pH) 0.1
For Use With (Application) Hold function, auto-off, error messages, waterproof, dustproof and floatable
Length (Metric) 16.5cm
Temperature (English) Operating 32° to 122°F
Temperature (Metric) Operating 0 to 50°C
Warranty 1 Year

  • Microprocessor-based, with accurate readings to 0.1pH
  • Replaceable electrode (S65290 or NC9830226) lets you use Testr body many times over, save money
  • Unique double junction design and extra reference gel volume for significantly longer electrode life, especially in harsh applications
  • Automatic temperature compensation
  • Push-button three-point calibration
  • Auto buffer recognition: selectable buffer sets
  • Hold function; auto-off; error messages
  • Waterproof, dustproof and floatable; certifiable to IP67 standards
Meter Only package includes meter, electrode, four 1.5V batteries, instructions.
Meter Kit package includes meter; electrode; two each of pH 4.01, 7.00, 10.00 buffers and rinse solution in single-use pouches; sample jar, batteries, instructions and hard plastic carrying case.

Ordering Information
Meter dimensions: 6-1/2 in. × 1-1/2 in. (16.5 × 3.8cm). Shipping weight (meter only):
4.5 oz. (125g).

For replacement electrodes, order Cat. No. NC9830226 (8 prongs) for Cat. No. 13-200-261. To confirm you are ordering the correct electrode, please call your Fisher Scientific Customer Service Representative.

One-year warranty.