missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oakton™ Turbidimeter

Waterproof and dustproof with IP67 rated housing that floats

Manufacturer:  Oakton™ 3563500

Catalog No. 13-300-251



Description

Description

  • Four-point, push button calibration for full-range accuracy
  • Auto-ranging from 0 to 1000 NTU
  • Portable and durable
  • Light weight
  • For use in field testing and quality control
  • Repeatability and accuracy
  • Resolution as low as 0.01 NTU

Specifications

Specifications

±2%, ±3%
Ideal for monitoring turbidity in chemical, food and industrial applications
0 to 1000 NTU
Portable Turbidity Meter
AAA Alkaline
1.8 in.
6.1 in.
67
6.8cm
6.1 x 2.7 x 1.8 in. (15.5 x 6.8 x 4.6cm)
Four primary standards, three empty cuvettes with light shield caps, silicone oil, lint-free cloth, 4 AAA batteries, instructions and hard carrying case
Infrared LED
0.01, 0.1, 1 NTU
LED
4.6cm
15.5cm
2.7 in.
SDS
Documents