Oakton™ Testr CTS Pocket Cond/TDS/Salinity Tester
These CTS testers make measuring water quality easier and faster. Three-in-one tool measures conductivity, TDS, and salinity
$380.81 - $380.81
Specifications
Backlight Option Yes
Battery Life 150+ hours
Battery Type 4 AAA Alkaline
Calibration Points 3
Channels 1
View More Specs
Includes: Protective plastic storage case, lanyard, and 4 AAA batteries.
Products 2
Catalog Number Mfr. No. Probe Type Price Quantity  
Catalog Number Mfr. No. Probe Type Price Quantity  
11-747-331
View Documents
Oakton™
WD3563455
Pin style integral conductivity cell
Each for $380.81
11-747-332
View Documents
Oakton™
3563465
Cup style integral conductivity cell
Each for $380.81
Description
Description
  • Redesigned leakproof cap functions as a sample cup and stabilizing base to stand the meter upright
  • Automatic temperature compensation ensures accurate reading in varying environments
  • Easy-to-navigate interface for quick setup and calibration
Warranty
1 Year
Specifications
Specifications
Yes
4 AAA Alkaline
1
Water quality testing, buffer and reagent production, electrochemistry
Conductvity, TDS, Salinity, Temperature
Checkmark indicator
Automatic
150+ hours
3
Graphics, Dot Matrix 80 x 100 pixel
IP 67
Hold Data
5°C to 45°C
0°C to 60°C