Oakton™ TDS 6+ Portable Meters
Set up function allows to customize automatic temperature compensation, cell constant and TDS conversion factor for better accuracy
Manufacturer:  Oakton™ 3560420
 View more versions of this product
Catalog No. 15-500-105
Includes: TDS 6+ Meter: Conductivity/TDS temperature probe (K=1), Protective rubber boot w/ stand, Batteries
TDS 6+ Meter Kit: Conductivity/TDS temperature probe (K=1), Protective rubber boot, Calibration solutions and sample bottles, Hard carrying case, Batteries
Description
Description
TDS 6+ Meter
  • Measures total dissolved salts (TDS)
  • High accuracy in changing temperature conditions
  • With temperature readout and automatic temperature compensation
  • One-point calibration is required for measuring all the five ranges
  • Both automatic and manual modes available for calibration
  • Easy push-button operation with hold and auto-off functions
  • Protective boot protects meter from drops and dings
  • Available with built-in stand
Specifications
Specifications
Conductivity/TDS temperature probe (K=1), protective Rubber boot with stand, batteries, built in ATCC
-10° to 110°C
Meter
0 to 10, 10 to 100, 100 to 1000ppm; 1 to 10, 10 to 100, up to 200ppt
-50° to 230°F
±0.5°C
±1% FS
0.01/0.1/1ppm, 0.01/0.1ppt
Documents